Läs mer
Etomoxir sodium salt, MedChemExpress
Produktkod. 16468068
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16468068 10 mg -
16410538 5 mg 5mg
16453987 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16468068 Leverantör Medchem Express Leverantörsnummer HY50202A
MedChemExpress Etomoxir((R)-(+)-Etomoxir) sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.
Etomoxir((R)-(+)-Etomoxir) sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.
Specifikationer
Kemiskt namn eller material Etomoxir sodium salt
CAS 828934-41-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C15H18ClNaO4
Kvantitet 10 mg
Synonym (R)-(+)-Etomoxir sodium salt
Löslighetsinformation DMSO : 100 mg/mL (311.78 mM; Need ultrasonic) ∣H2O : 5 mg/mL (15.59 mM; Need ultrasonic)
LEDER ClC1=CC=C(OCCCCCC[C@@]2(CO2)C(O[Na])=O)C=C1
Molekylvikt (g/mol) 320.74
Formel vikt 320.74
Procent renhet 98.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death