Läs mer
Etoricoxib N1’-Oxide, TRC
Produktkod. 30009033
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009033 100 mg
18090610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE934115100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 325855-74-1
Molekylformel C18 H15 Cl N2 O3 S
Molekylvikt (g/mol) 374.84
Synonym 5-Chloro-6'-methyl-3-[4-(methylsulfonyl)phenyl]-2,3'-bipyridine 1'-Oxide, Etoricoxib N1'-Oxide, 2,3'-Bipyridine, 5-chloro-6'-methyl-3-[4-(methylsulfonyl)phenyl]-, 1'-oxide (9CI, ACI)
IUPAC-namn 5-chloro-2-(6-methyl-1-oxidopyridin-1-ium-3-yl)-3-(4-methylsulfonylphenyl)pyridine
LEDER Cc1ccc(c[n+]1[O-])c2ncc(Cl)cc2c3ccc(cc3)S(=O)(=O)C
Specifikationer
Kemiskt namn eller material Etoricoxib N1'-Oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H15ClN2O3S/c1-12-3-4-14(11-21(12)22)18-17(9-15(19)10-20-18)13-5-7-16(8-6-13)25(2,23)24/h3-11H,1-2H3
Formel vikt 374.0492
Procent renhet >95%