Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Etrimfos, technical mixture |
| CAS | 38260-54-7 |
| CAS Min % | ≥ 50% |
| CAS Max % | <70% |
| Densitet | 1.200 g/cm3 (20°C) |
| Beilstein | 7136389 |
| Färg | Colorless |
| Smältpunkt | -3.35°C |
| Synonym | O-6-ethoxy-2-ethylpyrimidin-4-yl O,O-dimethyl phosphorothioate |
| Molekylvikt (g/mol) | 292.29 g/mol |
| Visa mer |