missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ ETV1 Monoclonal Antibody (1C8B6)
GREENER_CHOICE

Produktkod. 11375433
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11375433 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11375433 Leverantör Invitrogen™ Leverantörsnummer MA515461

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

MA5-15461 targets ETV1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15461 immunogen is purified recombinant fragment of ETV1 (aa1-191) expressed in E. Coli..

ETV1: ets variant gene 1. Several members of the Ets gene family encode sequence-specific DNA binding proteins that recognize DNA sequences with a centrally located 5'-GGAA-3' element. All of the Ets proteins recognize the same central core sequence but each protein interacts with unique sequences that flank this core. ER81 (also designated ETV1) binds the motif 5'-CGGAA/T-3'. ER81 is highly expressed in brain, testis, lung and heart. ER81 is also moderately expressed in spleen, pancreas, colon and small intestine. During development, ER81 displays a unique expression patterns which suggests that this transcriptional factor might play an important role in organogenesis. ERK-1 activates ER81 transcriptional activity while MAPKAP kinase 2 inhibits ER81.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen ETV1
Användningsområden ELISA, Western Blot
Klassificering Monoclonal
Klona 1C8B6
Koncentration Conc. Not Determined
Konjugera Unconjugated
Formulering ascites with 0.03% sodium azide
Gen Etv1
Genaccessionsnr. P50549
Gene Alias DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein
Gensymboler Etv1
Värd art Mouse
Immunogen Purified recombinant fragment of ETV1 (aa1-191) expressed in E. Coli.
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2115
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.