Läs mer
Invitrogen™ ETV3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140147
Beskrivning
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SSRFHFPPLD THSPTNDVQP GRFSASSLTA SGQESSNGTD RKTELSELED GSAADWRRGV DPVSSRNAIG GGGIGHQKR Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 75%.
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
Specifikationer
| ETV3 | |
| Polyclonal | |
| Unconjugated | |
| ETV3 | |
| AI414410; bA110J1.4; ETS domain transcriptional repressor PE1; ETS translocation variant 3; ETS variant 3; ets variant gene 3; ets variant gene 3, ETS family transcriptional repressor; ETS-domain protein; ETS-domain transcriptional repressor; Etv3; Mets; Mitogenic Ets transcriptional suppressor; mitogenic Ets transcriptional suppressor METS; Pe1; PE-1; SH2 domain containing phosphatase anchor protein 1; Spap1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41162 | |
| ETV3 | |
| Recombinant protein corresponding to Human ETV3. Recombinant protein control fragment (Product #RP-109328). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.