Läs mer
Eugenol Acetate, TRC
Produktkod. 30375614
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30375614 10 g
1 options
Produktkod. 30375614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE93865010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetyl Eugenol
CAS 93-28-7
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H14 O3
Kvantitet 10 g
Synonym Eugenol acetate (6CI), Phenol, 2-methoxy-4-(2-propenyl)-, acetate (9CI), Phenol, 4-allyl-2-methoxy-, acetate (7CI,8CI), 2-Methoxy-4-(2-propenyl)phenol acetate, 4-Allyl-2-methoxyphenyl acetate, Aceteugenol, Acetyleugenol, Eugenyl acetate, NSC 1242, O-Acetyleugenol, Phenol, 2-methoxy-4-(2-propen-1-yl)-, 1-acetate
InChI formel InChI=1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3
LEDER COc1cc(CC=C)ccc1OC(=O)C
IUPAC-namn (2-methoxy-4-prop-2-enylphenyl) acetate
Molekylvikt (g/mol) 206.24
Formel vikt 206.0943