missing translation for 'onlineSavingsMsg'
Learn More

Europium solution for ICP 10g/L HNO3 matrix, Fisher chemical™

Catalog Number p-7083914
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10±0.02g/L
Packaging Bottle
Quantity 100 mL
Type Mass Spectrometry Standard
Analyte or Component Names Eu
Solvent or Matrix HNO3
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'