Läs mer
Euscaphic Acid, TRC
Produktkod. 30378566
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30378566 5 mg
1 options
Produktkod. 30378566 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9395005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Euscaphic Acid
CAS 53155-25-2
Molekylformel C30H48O5
Kvantitet 5 mg
Synonym (2α,3α)-2,3,19-Trihydroxyurs-12-en-28-oic Acid, 2α,3α,19-Trihydroxy-12-en-28-ursolic Acid, 2α,3α,19α-Trihydroxyurs-12-en-28-oic Acid, Acuminatic Acid, Euscophic Acid, Euscapic Acid, Jacarandic Acid
InChI formel InChI=1S/C30H48O5/c1-17-10-13-30(24(33)34)15-14-27(5)18(22(30)29(17,7)35)8-9-21-26(4)16-19(31)23(32)25(2,3)20(26)11-12-28(21,27)6/h8,17,19-23,31-32,35H,9-16H2,1-7H3,(H,33,34)/t17-,19-,20+,21-,22-,23-,26+,27-,28-,29-,30+/m1/s1
LEDER C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@H]([C@H](C(C)(C)[C@@H]5CC[C@]43C)O)O)[C@@H]2[C@]1(C)O)C(=O)O
Molekylvikt (g/mol) 488.7
Formel vikt 488.3502