Learn More
Thermo Scientific™ Eutech PC700 Multiparameter
pH / mV-Redox-ORP/ Conductivity/ Total Dissolved Solids/ Temperature
The multi-parameter Eutech PC 700 measures pH, ORP, conductivity and temperature with a single meter. User-friendly with advanced features and a large custom-size display – perfect for research work on a budget!
Brand: Thermo Scientific™ ECPC70040S
Additional Details : Weight : 1.49400kg
Description
- Large screen in a compact footprint
- Meter accepts pH and Redox electrodes,in addition to Conductivity/TDS
- Up to 5-point push button calibration
- Selectable cell constant
- Auto-ranging across 5 Conductivity ranges
- Easy interchangeability of probes
- Non-volatile memory holds up to 100 data points
- Integral electrode holder
Specifications
±0.01 | |
Automatic (84μS, 1413μS, 12.88 mS, 111.8mS ); maximum 1 per range; manual (5 points); maximum 1 per range | |
Automatic with supplied cell or manual 0.0 to 100ºC /32.0 to 212.0ºF, 0 to 80ºC /32.0 to 176.0ºF with supplied cell | |
.. to 100.0 ppt @ 0.5 fact (200.0 @ 1 factor) | |
0.0°C to 100.0°C | |
PC700 Multiparameter bench meter | |
±0.5°C, ±0.9°F | |
32.0°F to 212.0°F |
± 1% full scale | |
0.1, 1.0, 10.0 (selectable) | |
-2.00 to 16.00 | |
0.01 | |
With pH Electrode (ECFC7252101B), Conductivity/ATC Probe CONSEN9501D), Integral Electrode Holder & 100/240 VAC Adapter | |
0.01/0.1/1μS; 0.01/ 0.1mS | |
Automatic with supplied cell or manual | |
0.1ºC/0.1 ºF |