Learn More
Thermo Scientific™ Eutech™ Thermocouple Penetration Probe, Type K ECTPPENK01M
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications.
Brand: Thermo Scientific™ 01X220102
Additional Details : Weight : 0.10000kg
Description
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications. The Eutech™ Thermocouple Penetration Probe, Type K ECTPPENK01M is an ungrounded, penetration probe used for penetrating meat, plastic & semi-soft materials.
Features:
- Parameter: Temperature
- Dimension: 120 x 3.2 mm
Specifications
-50°C to +700°C | |
Eutech meters: EcoScan Temp JKT, Temp 300, Temp 10K | |
3.2 mm | |
Miniature Plug |
Temperature Probe | |
Type K, Penetration probe (for penetrating meat, plastic & semi-soft materials), ungrounded | |
120 mm | |
1 m |