Learn More
Haldenwanger™ Evaporating Capsules
Use these dishes for a variety of applications. Haldenwanger™ Evaporating Capsules feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ 130/6
Additional Details : Weight : 0.13500kg
Description
- Available in a variety of sizes
- Sizes up to 175mL glazed except rim
- 1,000mL and larger are glazed except outside base
- Complies with DIN VDE 0335, type C 100
Specifications
Round | |
Porcelain | |
Evaporating Capsule | |
52 mm |
270 mL | |
Round | |
Laboratories | |
110 mm |