Läs mer
Everolimus Isomer C (>60%, contains everolimus), TRC
Produktkod. 30137101
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137101 25 mg
30135820 1 mg
30007188 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE94542525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 908340-97-6
Molekylformel C53 H83 N O14
Molekylvikt (g/mol) 958.22
Synonym Rapamycin, 9,14-deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-42-O-(2-hydroxyethyl)-14-oxo- (9CI), Everolimus Isomer C, 9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-42-O-(2-hydroxyethyl)-14-oxorapamycin, Everolimus oxepane isomer, (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,28R,34aS)-9,10,12,13,14,21,22,23,24,25,26,28,32,33,34,34a-Hexadecahydro-9,28-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,28-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,27,29(4H,6H,31H)-pentone
LEDER CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)C(=O)[C@@](O)(O3)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1OCCO
Specifikationer
Kemiskt namn eller material 9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-42-O-(2-hydroxyethyl)-14-oxorapamycin
Kvantitet 25 mg
InChI formel InChI=1S/C53H83NO14/c1-32-16-12-11-13-17-33(2)44(63-8)30-40-21-19-34(3)50(59)53(62,68-40)52(61)54-23-15-14-18-41(54)51(60)67-45(36(5)28-39-20-22-43(66-25-24-55)46(29-39)64-9)31-42(56)35(4)27-38(7)48(58)49(65-10)47(57)37(6)26-32/h11-13,16-17,27,32,34-37,39-41,43-46,48-49,55,58,62H,14-15,18-26,28-31H2,1-10H3/b13-11+,16-12+,33-17+,38-27+/t32-,34-,35?,36-,37-,39+,40+,41+,43-,44+,45+,46-,48-,49+,53-/m1/s1
Formel vikt 957.5814