missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Evofosfamide, MedChemExpress
Beskrivning
Evofosfamide (TH-302) is a hypoxia-activated prodrug with IC50 of 10 μM and 1000 μM in hypoxia (N2) and normoxia (21% O2), respectively.
Specifications
Specifications
| Kemiskt namn eller material | Evofosfamide |
| CAS | 918633-87-1 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C9H16Br2N5O4P |
| Kvantitet | 50 mg |
| Synonym | TH-302 |
| Löslighetsinformation | DMSO : 94 mg/mL (209.34 mM; Need ultrasonic and warming) |
| LEDER | O=P(NCCBr)(OCC1=CN=C(N1C)[N+]([O-])=O)NCCBr |
| Molekylvikt (g/mol) | 449.04 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?