Läs mer
Thermo Scientific™ Evolution™ 201/220 UV-Visible Spectrophotometers
Produktkod. p-4532046 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Produktlinje:
Evolution 220
Typ:
Datorstyrningsspecifikation
missing translation for 'unitSize'
Varje
Product Code. 10254038
missing translation for 'mfr': Thermo Scientific™ 840210600
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Empower your analysis and bring versatility and productivity to your lab with the new Thermo Scientific™ Evolution 201 and 220 UV-Vis systems.
New sales of this model have been discontinued. Please see the newest models: the Evolution One/One Plus Spectrophotometers.
For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution 201 and 220 systems work with you to perform experiments the way you want - for the results you need.
INSIGHT Software—Sophistication Simplified
  • Personalize your home screen for easy navigation and convenience
  • Workbook tools save time and ensure accurate results
  • CUE scripting allows creation of customized interface and simplified workflow
Optimized Engineering Delivers Ultimate Performance
  • Double-beam geometry with 1nm spectral bandwidth for precision measurements
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Electronic triggering provides the highest level of accuracy for rapid-mixing kinetics measurements
  • Precision monochromator drive delivers fast scanning data collection and wavelength accuracy
Powerful Convenience at your Fingertips
  • Color, touch screen provides instrument control using built-in computer
  • Integrated keypad allows communication with INSIGHT software
  • Choice of Local or PC configurations provides flexibility and convenience
Accessories for all your Sampling Needs
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements
Assurance of System Performance
  • The Thermo Scientific UV Validator package provides support for system qualification and validation activities
  • Documentation facilitates compliance of your system with the requirements of FDA,GxP, ISO 9001:2000 and ISPE 2001 guidelines and regulations
  • Mercury Lamp accessory is available to perform instrument calibration
  • Thermo Scientific INSIGHT security software provides complete tools to meet 21 CFR Part 11 requirements
TRUSTED_SUSTAINABILITY
Specifications
Baslinjeplanhet ±0.0010 A, 200 to 800 nm, 1.0 nm SBW, smoothing
Beskrivning Evolution 220 UV-Vis Spectrophotometer, Computer Control, includes Euro/US/UK power cords
Detektortyp Dual Silicon Photodiodes
Visa No Display
Mått (L x B x H) 62.2 x 48.6 x 27.9 cm (24 x 19 x 11 in.)
Produktlinje Evolution 220
Spektral bandbredd Variable: 1.0 nm, 2.0 nm
Typ Datorstyrningsspecifikation
Våglängdsområde 190 to 1100 nm
Drift <0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hr. warmup
Elektriska krav 100/240 V, 50/60 Hz selected automatically, 150 W maximum
Inkluderar EU/US/UK power cords
Knappsats Sealed membrane
Lampans livslängd 7 years typical
Buller 0A: <0.00015A;
1A: <0.00025A;
2A: <0.00080A;
260 nm, 1nm SBW, RMS
Optisk design AFBG Microcell optimized, AFBG Fiber optic optimized, AFBG Materials optimized
Farmakopéöverensstämmelsetestning (Guaranteed Performance Specifications)
Resolution (toluene in Hexane): ≥1.8A
Photometric Accuracy (60 mg/L K2Cr2O7): ±0.010A
Stray Light: ≤1%T at 198 nm: KCI; ≤ to 0.05 %AT at 220nm: Nal, Kl
Wavelength Accuracy: ±0.5 nm 541.9, 546.1 nm Hg emission lines, ±0.8 nm full range
Wavelength Repeatability: ≤0.05 nm, repetitive scanning of 546.1nm Hg emission line
Fotometrisk noggrannhet 1 A: ±0.006A, 2 A: ±0.010 A, Measured at 440 nm
Fotometrisk display -0.3 to 4.0A
Fotometriskt område >3.5A
Fotometrisk repeterbarhet ±0.0002 A
Processor Microsoft Windows 7, Windows 8
Skanna ordningslägen Absorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs*Factor, Intensity
Skanningshastighet <1 to 6000 nm/min. (Variable)
Garanti 3 Years
Våglängdsnoggrannhet ±0.5 nm (541.9, 546.1 nm mercury lines), ±0.8 nm (full range 190 to 1100 nm)
Våglängdsdataintervall 10, 5, 2, 1.0, 0.5, 0.2, 0.1 nm
Våglängds repeterbarhet ≤0.05 nm (546.11 nm mercury line, SD of 10 measurements)
Vikt (engelska) 32 lb.
Vikt (metrisk) 14.4 kg