Läs mer
Safety First Aid Evolution HypaSoothe Burns Kit, Small
Beskrivning
- The kit is delivered in a durable Evolution case (15 x 23 x 8 cm) capable of protecting its contents from dust and damp.
- It is easy to identify with its bright red Coloring, flame logo and Burns Kit label.
- It can be wall mounted with the help of a bracket for ease of storage.
- Bracket sold separately
- Kit Contents:
- 1x First Aid Guidance Leaflet
- 2x HypaSoothe Burn Dressings, 10 x 10 cm
- 3x HypaSoothe Emergency Burn Gel Sachets, 6 g
- 12x HypaBand Safety Pins
- 2x HypaBand Conforming Bandages
- 2x HypaTouch Disposable Gloves (Pairs)
Specifikationer
Specifikationer
| Produkttyp | First Aid Kit |
| Material | Plastic |
| Sterilitet | Sterile |