Läs mer
Safety First Aid Evolution Plus Critical Injury Bleed Control Kit, Medium
Beskrivning
- It is ideal for the first aider to take directly to the site of the incident.
- The bleed control kit is perfect for high-risk industries, such as construction, industrial and agricultural, where there is a risk of severe injury.
- Having a tourniquet and haemostatic gauze included in a workplace first aid kit ensures that any injury with critical, life-threatening bleeding can be treated.
- Having our Evolution Plus Critical Injury Bleed Control Kit in the workplace is also essential for those that are in more isolated locations, where it may take longer for emergency services to reach the casualty.
- On average, an ambulance arrives within 9 minutes in the UK, but this will depend on availability and the location they are travelling to, which can impact the injured parties’ chance of survival.
- With the supplies provided in this kit, staff are able to treat any injuries that may have occurred and prolong the casualty's life until paramedics arrive.
- With the implementation of Martyn’s Law, a legislation aimed at improving public safety through better preparedness against terrorist threats, businesses and public venues will be expected to take proactive steps in preparing for potential mass casualty incidents.
- Designed for use in high-stress, emergency scenarios such as terror attacks, violent assaults, or serious accidents, the Critical Injury Bleed Control Kit equips staff with the essential tools to provide immediate, potentially life-saving care before emergency services arrive.
- Meaning that this kit can play a vital role in fulfilling these responsibilities.
- By having a bleed control kit on site, your venue demonstrates a clear commitment to safety and compliance.
- It ensures you are prepared to treat individuals suffering from catastrophic injuries, including severe bleeding-one of the most common and preventable causes of death in such incidents.
- Beyond regulatory compliance, these kits provide staff with confidence and peace of mind, knowing they have the means to respond effectively in a crisis.
- They can also help reduce the potential human and reputational impact of a mass casualty event on your organisation.
- In the event of a life-threatening injury, particularly one involving severe bleeding, every second counts.
- A critical injury kit is specifically designed to equip first responders and bystanders with the tools needed to manage catastrophic blood loss before emergency services arrive.
- One of the leading causes of preventable death in trauma situations is uncontrolled bleeding.
- This kit includes advanced medical supplies such as a tourniquet to restrict blood flow from a limb and haemostatic dressings that accelerate clotting at the wound site.
- These items are proven to significantly increase a casualty’s chances of survival by controlling bleeding quickly and effectively.
- Having a Critical Injury Kit on hand can make the difference between life and death, whether it is in a workplace, public venue, or remote location.
- 2 x HypaCover Trauma Dressings (Large) - These dressings absorb larger volumes of exudate than normal dressings, allowing pressure to be maintained with the dressings needing to be removed less frequently.
- 2 x HypaTouch Nitrile Gloves - Having gloves reduces the risk of infection both to the wound and to the first aider from blood borne viruses.
- 2 x HypaStop Haemostatic Gauze (10 x 10 cm) - These dressings are specially treated wound dressings designed to rapidly stop bleeding, especially in severe or life-threatening situations. They contain agents that accelerate the blood clotting process when applied directly to a wound.
- 1 x HypaGuard Foil Blanket - Ideal for casualties in shock or suffering from hypothermia.
- 1 x Emergency Tourniquet - Apply the tourniquet above the wound and tighten to stop blood flow, thus stopping the life-threatening bleeding.
- 1 x Clothing Cutters - Removes clothing that is obstructing access to the wound or preventing first aid treatment from taking place.
- 1 x First Aid Guidance Leaflet - Offers basic first aid instructions, enabling even untrained individuals to effectively use the kit's supplies.
Specifikationer
Specifikationer
| Produkttyp | First Aid Kit |
| Material | Plastic |
| Sterilitet | Sterile |