Läs mer
Invitrogen™ EVOS™ Light Cubes
Klicka för att se tillgängliga alternativ
Kvantitet:
1 Each
Förpackningsstorlek:
Varje
Beskrivning
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifikationer
Specifikationer
| Typ | LED Light Cube |
| Beskrivning | EVOS™ Qdot™ WB 525-800 Light Cube, LED, Unique, For Use With EVOS™ FL Auto Imaging System, EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, Fluorophores with Excitation and Emission Maxima Near 445 and 525 to 800nm |
| För användning med (utrustning) | EVOS™ FL Imaging System, EVOS™ FL Auto 1 and 2 Imaging System, EVOS™ M5000/M7000 Imaging System, Countess™ II FL Automated Cell Counter |
| Kvantitet | 1 Each |