Learn More
EVOS™ EVOS™ Onstage Master Plate
Allows user to utilize different types of vessels for imaging
Brand: EVOS™ AMEPVH035
Additional Details : Weight : 2.00000kg
Description
- Part of EVOS Onstage Incubator and holds EVOS Onstage Vessel Holder
- Is an environmental chamber designed specifically for use with EVOS FL Auto Imaging System
- Accommodates various types of cell culture vessels
- Provides flexibility to extended time-lapse imaging experiments under precisely controlled environmental conditions
Specifications
Master Plate | |
EVOS™ FL Auto Imaging System | |
1 Ea. |
EVOS | |
Onstage Vessel |
For Research Use Only. Not for use in diagnostic procedures.