Learn More
Fisherbrand™ Excursion Trac Thermometer
Use this thermometer for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Fisherbrand™ Traceable™ Excursion Trac Thermometers feature user-defined time intervals.
Brand: Fisherbrand™
Additional Details : Weight : 0.39600kg
Description
- Meets current CDC requirements for vaccine storage and monitoring
- Rolling memory structure, maintains most recent 525,600 temperature observations
- Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
- Stores up to 10 unique alarm events
- Memory can be cleared on unit after downloading data to USB stick
- Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
- Status indicators - Low battery, memory full, USB data transfer and active alarm state