missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20138-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
EXD2 | |
Polyclonal | |
Unconjugated | |
EXD2 | |
C14orf114, EXD2, EXDL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362759, 55218, 97827 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VEG4, Q9NVH0 | |
Exd2 | |
EXD2 Fusion Protein Ag14090 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |