Läs mer
Exenatide Acetate, TRC
Produktkod. 30404342
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30404342 5 mg
1 options
Produktkod. 30404342 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9573005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Exenatide Acetate
CAS 141732-76-5
Molekylformel C186H286N50O62S
Kvantitet 5 mg
Synonym Exendin-4
LEDER NC([C@H](CO)NN1CCC[C@H]1C(C([C@H]2N(CCC2)C([C@H]3N(CCC3)C([C@H](NC(CNC([C@H](CO)NC([C@H](CO)NC([C@H]4N(CCC4)C(CNC(CNC([C@H](CC(N)=O)NC([C@H](CCCCN)NC([C@H](CC(C)C)NC([C@@H](NC([C@H](CCC(O)=O)NN[C@H](C(C([C@@H](NC([C@H](CC(C)C)NC([C@H](CCCNC(N)=N)NC([C@H](C(C)C)NC([C@H](NC([C@H](CCC(O)=O)NC([C@H](CCC(O)=O)NC([C@H](CCC(O)=O)NC([C@@H](NC([C@H](CCC(N)=O)NC([C@H](CCCCN)NC([C@H](CO)NC([C@H](CC(C)C)NC([C@@H](NC([C@H](CO)NC([C@H]([C@H](O)C)NC([C@@H](NC([C@H]([C@H](O)C)NC(CNN[C@@H](CCC(O)=O)C(NCC(C([C@@H](N[H])CC5=CN=CN5)=O)=O)=O)=O)=O)CC6=CC=CC=C6)=O)=O)=O)CC(O)=O)=O)=O)=O)=O)=O)CCSC)=O)=O)=O)=O)C)=O)=O)=O)=O)CC7=CC=CC=C7)=O)=O)[C@@H](C)CC)=O)CC8=CNC9=C8C=CC=C9)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)C)=O)=O)=O)=O)=O.CC(O)=O
Molekylvikt (g/mol) 4246.62