Läs mer
3M™ Exhalation Valve
Produktkod. 15739657 Handla allt 3M Produkter
Change view
:
Color: Black
Produktkod.
15739657 Color: Black
1 options
Produktkod. 15739657 Leverantör 3M™ Leverantörsnummer 7000029732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
3M™ Exhalation Valve is suitable for use with 3M™ Reusable Half Mask 7000 Series and the 3M™ Centre Adapter 6864 on the 3M™ Reusable Full Face Mask 6000 Series.
  • Replacement exhalation valve for half face respirator and also used with the center adapter on full facepiece respirators
  • Replace damaged parts to ensure correct levels of protection and comfort
  • For use with 3M™ 7000 Series Half Face Respirator and Centre Adapter 6864 on the 3M™ 600 Full Facepiece Respirators 6000 Series
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad CE Approved
Färg Black
Produkttyp Exhalation Valve
För användning med (utrustning) 7500 Series Half Mask