missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXO1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | EXO1 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835834
|
Proteintech
16253-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825834
|
Proteintech
16253-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.Specifications
EXO1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9QZ11, Q9UQ84 | |
26909, 9156 | |
EXO1 Fusion Protein Ag9253 | |
Primary | |
-20°C | |
EXO1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI | |
EXO1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |