missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXO1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16253-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.Specifications
EXO1 | |
Polyclonal | |
Unconjugated | |
EXO1 | |
EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26909, 9156 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QZ11, Q9UQ84 | |
EXO1 | |
EXO1 Fusion Protein Ag9253 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |