missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | EXOC5 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877524
|
Proteintech
17593-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867524
|
Proteintech
17593-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.Specifications
EXOC5 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O00471, P97878, Q3TPX4 | |
105504, 10640, 60627 | |
EXOC5 Fusion Protein Ag11524 | |
Primary | |
-20°C | |
EXOC5 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EXOC5, exocyst complex component 5, HSEC10, PRO1912, SEC10, SEC10L1, SEC10P | |
Exoc5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |