Läs mer
Tocris Bioscience™ EXP 3174
Potent and selective AT1 receptor antagonist
Brand: Tocris Bioscience™ 5728/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
EXP 3174 is a potent and selective noncompetitive AT1 receptor antagonist (IC50 = 37 nM). Reduces blood pressure in a rat model. Active metabolite of losartan potassium (Cat.No. 3798).Specifikationer
| 2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid | |
| 124750-92-1 | |
| 50 mg |