Learn More
BINDER™ External Gas Tank Changer
For Use With: series C, CB
Brand: BINDER™ 8012-0408
Additional Details : Weight : 2.83000kg
Description
- External gas tank changer
- For either CO2 or N2
- With audible and visual alarm
- Zero-voltage alarm output
Specifications
Gas tank changer - external, BINDER Gas Supply Service, for connecting 2 ga tanks (CO2, N2 or O2), with audible and visual alarms, as well as zero-voltage alarm outpμt | |
Accessory, Gas Tank Connector | |
Audible and visual alarm |
BINDER Chambers Series CB-S, CB, CBF | |
Gas Tank Connections |