Learn More
BINDER™ External Gas Tank Changer
For Use With: series C, CB
Brand:  BINDER™ 8012-0408
Additional Details : Weight : 2.83000kg
Product Code. 15641736
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • External gas tank changer
  • For either CO2 or N2
  • With audible and visual alarm
  • Zero-voltage alarm output
Specifications
Specifications
Gas tank changer - external, BINDER Gas Supply Service, for connecting 2 ga tanks (CO2, N2 or O2), with audible and visual alarms, as well as zero-voltage alarm outpμt
Accessory, Gas Tank Connector
Audible and visual alarm
BINDER Chambers Series CB-S, CB, CBF
Gas Tank Connections