missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EYA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22658-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.Specifications
EYA1 | |
Polyclonal | |
Unconjugated | |
EYA1 | |
BOP, BOR, EYA1, Eyes absent homolog 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14048, 2138 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97767, Q99502 | |
EYA1 | |
EYA1 Fusion Protein Ag18489 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |