Learn More
Ekastu Safety™ EPS Eye Wash Station
Product Code. 10758025
Change view
Click to view available options
Includes:
Pull-top Lid
Unit Size:
Each
Product Code. Includes Unit Size
10758025 Pull-top Lid Each
1 options
Product Code. 10758025 Supplier Ekastu Safety™ Supplier No. 277955
 Restricted Item
Beräknad leverans: 16-12-2026
Protects eye wash bottles from over exposure to warmth or cold. EKASTU Safety™ EPS Eye Wash Station indicates the temperature delta of water content ±19°C.
  • For direct availability in danger areas
  • Dust-proof with a pull-top lid
  • Protects eye wash bottles from warmth and cold and indicates the temperature delta of water content approx. 38°C (corresponds to approx. ±19°C). The eye wash station protects the contents up to five times better
  • Date of expiry can be stated on the lid
  • Easy installation
Specifications
For Use With (Application) Protects Eye Wash Bottles
Includes Pull-top Lid
Product Type Eye Wash Station