Learn More
Fisherbrand™ Eyepiece Pair for Magnum Series
Designed for use with Magnum Series microscopes. Fisherbrand™ Eyepiece Pairs are ergonomically designed for everyday use.
1945.00 SEK - 2905.00 SEK
Specifications
Type Eyepiece Pair
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Magnification Power Price Quantity & Availability  
Product Code Brand Magnification Power Price Quantity & Availability  
11552663
View Documents Promotion Details
Fisherbrand™
10X
1945.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
11592643
View Documents Promotion Details
Fisherbrand™
16X
2905.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
The trinocular versions allow connection to cameras whilst the planachromatic objectives provide true colour reproduction excluding spherical aberration.
The phase contrast models allow an efficient view of thin or translucent specimens where staining is difficult. Capable of Kohler illumination, accessories are also available for darkfield and polarisation enabling a wide range of applications. For comfort the head is inclined at 30° with interpupillary distance adjustment (Siedentopf type) of 55mm to 75mm. The quintuple reversed nosepiece revolves with click stop. With blue and green filter, 1 x spare bulb, 2 x spare fuses, immersion oil 10mL, soft rubber eyecups, dust cover and instruction manual.
Images may illustrate the microscope with Coolpix digital camera accessory: please note this is an optional extra and is NOT supplied as standard. For details, please contact Customer Service.
Specifications
Specifications
Eyepiece Pair
1 Ea.