Läs mer
EZ-Link NHS-PEG12-Biotin, TRC
Produktkod. 30482030
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482030 25 mg
30475890 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30482030 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN39401025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 365441-71-0
Molekylformel C19 H28 N4 O7 S
Synonym α-[3-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-3-oxopropyl]-ω-[2-[[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]ethoxy]poly(oxy-1,2-ethanediyl)
Specifikationer
Certifikat och efterlevnad ISO 9001
Molekylvikt (g/mol) 456.513