Läs mer
Ezetimibe Ketone, TRC
Produktkod. 17938404
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17938404 1 mg
17947704 5 mg
17977614 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17938404 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9750651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 191330-56-0
Molekylformel C24 H19 F2 N O3
Molekylvikt (g/mol) 407.41
Synonym 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)-, (3R,4S)-, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)-, (3R-trans)-, (3R,4S)-1-(4-Fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)azetidin-2-one
IUPAC-namn (3R,4S)-1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)azetidin-2-one
LEDER Oc1ccc(cc1)[C@@H]2[C@@H](CCC(=O)c3ccc(F)cc3)C(=O)N2c4ccc(F)cc4
Specifikationer
Kemiskt namn eller material Ezetimibe Ketone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C24H19F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21,23,28H,13-14H2/t21-,23-/m1/s1
Formel vikt 407.1333
Procent renhet >95%