Läs mer
EZM 2302, MedChemExpress
Produktkod. 16467127
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16467127 5 mg
16454408 10 mg
16450727 50 mg
16425306 100 mg
16423066 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16467127 Leverantör Medchem Express Leverantörsnummer HY111109
MedChemExpress EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM.
EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM.
Specifikationer
Kemiskt namn eller material EZM 2302
CAS 1628830-21-6
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C29H37ClN6O5
Kvantitet 5 mg
Löslighetsinformation DMSO : 100 mg/mL (170.91 mM; Need ultrasonic)
LEDER CNC[C@@H](O)COC1=CC=C(Cl)C(C2=NC(N3CC4(CCN(C(OC)=O)CC4)C3)=C(C)C(C5=C(C)ON=C5C)=N2)=C1
Molekylvikt (g/mol) 585.09
Formel vikt 585.09
Procent renhet 99.28%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death