missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, TRITC

Produktkod. 15440934
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μg
Förpackningsstorlek:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15440934 500 μg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15440934 Leverantör Invitrogen™ Leverantörsnummer A24511

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

F(ab')2-Donkey Polyclonal Secondary Antibody

Rehydrate with 0.55 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. 1 year from date of receipt. Prepare working dilution prior to use and then discard. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin mouse serum immunoglobulins, IgG from bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat or sheep.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Mouse IgG (H+L) Highly Cross-Adsorbed
Användningsområden Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera TRITC
Formulering PBS with 1% BSA and 0.05% sodium azide; pH 7.2
Värd art Donkey
Immunogen Gamma Immunoglobins Heavy and Light chains.
Reningsmetod Affinity Chromatography
Kvantitet 500 μg
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Mouse
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Produkttyp Secondary Antibody
Form Lyophilized
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.