Läs mer
R&D Systems™ F. keratolyticus Endo-beta-Galactosidase Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 8620-GH-050
Encompass
Produktkod. 15730274
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
Measured by its ability to cleave keratan sulfate. 50% cleavage of keratan sulfate (0.5 μg) is achieved using <5 ng rF.k. Endo-beta-galactosidase, as measured under the described conditions.
Specifikationer
Specifikationer
Supplied as a 0.2 µm filtered solution in Tris and NaCl.
50 µg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endo-beta-Galactosidase
Unconjugated
M.W. (Observed): 66 kDa, reducing conditions; M.W. (theoretical): 59 kDa
E. coli-derived f. keratoyticus Endo-beta-Galactosidase protein msyB (Thr2-Arg124) (N-terminus) Accession # P25738 GSENLYFQGHMHHHHHH F. keratolyticus Endo-beta-Galactosidase (Asp25-Leu422) (N-terminus) Accession # Q9ZG90 (C-terminus)
<1.0 EU / 1 µg of the protein by the LAL method.
Measured by its ability to cleave keratan sulfate. 50% cleavage of keratan sulfate (0.5μg) is achieved using <5 ng rF.k. Endo-beta-galactosidase, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.