missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
F4/80 Antibody (CI-A3-1), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB600-404UV
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifikationer
| F4/80 | |
| Monoclonal | |
| DyLight 350 | |
| ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
| Rat | |
| 160 kDa | |
| 0.125 mL | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 2015 | |
| Human, Mouse, C. elegans, E. coli | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CI-A3-1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ADGRE1 | |
| Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages. | |
| Store at 4C in the dark. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering