missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ F4/80 Monoclonal Antibody (BM8), Functional Grade, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

3320.00 SEK - 158800.00 SEK

Specifikationer

Antigen F4/80
Klona BM8
Koncentration 1 mg/mL
Klassificering Monoclonal
Konjugera Functional Grade
Se fler specifikationer

Produkter 4
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
15268627
Visa dokument
Invitrogen™
16-4801-85
500 μg
3320.00 SEK
500µg
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
17141252
Visa dokument
Invitrogen™
16-4801-38
5 mg
35060.00 SEK
5mg
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
17121252
Visa dokument
Affymetrix eBioscience
16-4801-025
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
17131252
Visa dokument
Affymetrix eBioscience
16-4801-050
50 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
Beskrivning

Beskrivning

The BM8 MAb reacts with mouse F4/80 antigen, an approximately 125 kDa transmembrane protein. The F4/80 antigen is expressed by a majority of mature macrophages and is the best marker for this population of cells. However, other cell types such as Langerhans cells and liver Kupffer cells have been reported to express this antigen. Expression of F4/80 commences during early myeloid development and is upregulated on all BM cells stimulated in vitro with M-CSF. It has been shown that some cytokines downregulate the expression of F4/80 resulting in lack of F4/80 antigen on a subpopulation of macrophages, especially in the lymphoid microenvironment in vivo.

F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Specifikationer

Specifikationer

F4/80
1 mg/mL
Functional Grade
Rat
Mouse
Q61549
IgG2a κ
Affinity Chromatography
Antibody
BM8
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
13733
Primary
4°C
ADGRE1
Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.