missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ FA2H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524728
This item is not returnable.
View return policy
Description
This antibody is predicted to react with non-human primate and rat based on sequence homology.
FA2H is a protein that catalyzes the synthesis of 2-hydroxysphingolipids, a subset of sphingolipids that contain 2-hydroxy fatty acids. Sphingolipids play roles in many cellular processes and their structural diversity arises from modification of the hydrophobic ceramide moiety, such as by 2-hydroxylation of the N-acyl chain, and the existence of many different head groups.
Specifications
| FA2H | |
| Polyclonal | |
| Unconjugated | |
| FA2H | |
| FA2H; FAAH; FAH1; Fatty acid 2-hydroxylase; fatty acid alpha-hydroxylase; fatty acid hydroxylase domain containing 1; Faxdc1; FLJ25287; G630055L08Rik; RGD1310347; SCS7; spastic paraplegia 35 (autosomal recessive); SPG35; WD repeat domain 59; Wdr59 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 338521, 79152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5MPP0, Q7L5A8 | |
| FA2H | |
| KLH conjugated synthetic peptide between 121-148 amino acids from the central region of human FA2H. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction