Läs mer
Bolle Safety Faceguard for Spectrum Safety Goggles
GREENER_CHOICE
Produktkod. 31810779 Handla allt Bollé Safety Produkter
Change view
Klicka för att se tillgängliga alternativ
Ram färg:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Ram färg
31810779 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 31810779 Leverantör Bolle Safety Leverantörsnummer SPECTN00W
 Begränsad artikel
Beräknad leverans: 04-09-2026
Lägg i varukorgen
Lägg i varukorgen
Enhances facial protection against impacts and debris, improving safety coverage while maintaining comfort in demanding environments.
  • Extended facial protection: Adds an extra layer of safety by protecting the lower face from impacts, particles, and debris.
  • Designed for SPECTRUM compatibility: Specifically developed for seamless integration with SPECTRUM safety goggles (regular size).
  • Improved coverage: Enhances overall PPE protection by extending the protective area beyond the eyes.
  • Maintains comfort: Lightweight design ensures user comfort without compromising protection during extended wear.
  • Secure and stable fit: Attaches firmly to goggles to remain in place during movement and demanding tasks.
  • Supports compliance: Increased protection and comfort encourage consistent use in high-risk environments.
  • Durable construction: Built to withstand daily industrial use, helping reduce replacement frequency.
  • Ideal for high-exposure environments: Suitable for industries such as construction, manufacturing, and maintenance where additional face protection is required.
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad EN ISO 16321-1
Linsbeläggning Uncoated
Färg Black
Linsfärg Clear
Produkttyp Safety Goggles
Ram färg Blue
Material Polycarbonate
Disponibel Reusable
Linsmaterial Plastic
Rammaterial Polycarbonate
UV-skydd None
Ramtyp Frameless
Autoklaveringsbar Not Autoclavable