Learn More
Memmert™ Factory Calibration Certificate For 3 Temperatures (37/52/70°C), For I30
Memmert™ Calibration Certificate with a single measuring point is for three temperatures.
Brand:  Memmert™ I30D00126
Additional Details : Weight : 0.01100kg
Product Code. 16375387
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Works calibration certificate for 3 temperatures: +37°C, +52°C, +70°C
  • Standard works calibration certificate (measuring point chamber centre) at +80°C
Specifications
Specifications
Measuring point chamber centre
Standard Works Calibration Certificate
37°C, 52°C, 70°C
Certificate for 3 temperatures