missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FAF2 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895824
|
Proteintech
16251-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16885824
|
Proteintech
16251-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Specifications
FAF2 | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q3TDN2, Q5BK32, Q96CS3 | |
23197, 291000, 76577 | |
FAF2 Fusion Protein Ag9236 | |
Primary | |
-20°C | |
FAF2 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
Faf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |