missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16251-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Specifications
FAF2 | |
Polyclonal | |
Unconjugated | |
FAF2 | |
ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23197, 291000, 76577 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TDN2, Q5BK32, Q96CS3 | |
Faf2 | |
FAF2 Fusion Protein Ag9236 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |