Läs mer
FAICAR (>80%), TRC
Produktkod. 30009222
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009222 2.5 mg 2.5mg
30007599 1 mg -
30006395 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF1010652.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13018-54-7
Molekylformel C10H15N4O9P
Molekylvikt (g/mol) 366.22
Synonym 5-Formylamino-4-imidazolecarboxamide Ribonucleotide, 5-(Formylamino)-1-(5-O-phosphono-β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, 5-Formamido-1-β-D-ribofuranosyl-,5'-(dihydrogen phosphate)-imidazole-4-carboxamide
IUPAC-namn ((2R,3S,4R,5R)-5-(4-carbamoyl-5-formamido-1H-imidazol-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
LEDER O[C@H]1[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2NC=O)O[C@@H]1COP(O)(O)=O
Specifikationer
Kemiskt namn eller material ((2R,3S,4R,5R)-5-(4-carbamoyl-5-formamido-1H-imidazol-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
Kvantitet 2.5 mg
InChI formel InChI=1S/C10H15N4O9P/c11-8(18)5-9(13-3-15)14(2-12-5)10-7(17)6(16)4(23-10)1-22-24(19,20)21/h2-4,6-7,10,16-17H,1H2,(H2,11,18)(H,13,15)(H2,19,20,21)/t4-,6-,7-,10-/m1/s1
Formel vikt 366.06