missing translation for 'onlineSavingsMsg'
Läs mer

FAM101A Antibody, Novus Biologicals™

Produktkod. 18298042 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18298042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18298042 Leverantör Novus Biologicals Leverantörsnummer NBP258508

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

FAM101A Polyclonal specifically detects FAM101A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM101A
Användningsområden Immunocytochemistry, Immunofluorescence
Klassificering Polyclonal
Konjugera Unconjugated
Utspädning Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulering PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias family with sequence similarity 101, member A, FLJ44614
Gensymboler FAM101A
Värd art Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FFGESIKVNPEPTHEIRCNSEVKYASEKHFQDKVFYAPVPTITAYSETIVAAPNCTWRNYRSQLTLEPRPRALRFRSTTIIFPKHARSTFRTTLHCSLGRPSRWFTASVQLQLCQDP
Reningsmetod Affinity Purified
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 144347
Testspecificitet Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Målarter Human
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotyp IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.