missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM105A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25668-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FAM105A belongs to the FAM105 family. The exact function of FAM105A remains unknown.Specifications
FAM105A | |
Polyclonal | |
Unconjugated | |
FAM105A | |
FAM105A, NET20, Protein FAM105A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54491 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NUU6 | |
FAM105A | |
FAM105A Fusion Protein Ag22243 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |