Learn More
FAM110B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552543
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KVTSVKPLKA IPCSSSAPPL PPKPKIAAIA SMKSPEADPV EPACGVSRRP SLQRSKSDLS DRYFRVDADV ERFFNYCGLD PEELENLGME NFARANSDII SLNFRSASMI SSDC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
FAM110B Polyclonal antibody specifically detects FAM110B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FAM110B | |
Polyclonal | |
Unconjugated | |
FAM110B | |
1700012H17Rik; 5031405P22; AW049855; C8orf72; FAM110B; family with sequence similarity 110 member B; family with sequence similarity 110, member B; protein FAM110B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
242297, 500400, 90362 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5BJX5, Q8C739, Q8TC76 | |
FAM110B | |
Recombinant protein corresponding to Human FAM110B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only