missing translation for 'onlineSavingsMsg'
Learn More

FAM133A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24574-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883055

  • 1355.00 SEK / 20µL
Estimated Shipment: 23-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FAM133A is a protein coding gene.
Specifications

Specifications

FAM133A
Polyclonal
Unconjugated
FAM133A
FAM133A, Protein FAM133A
Rabbit
Antigen Affinity Chromatography
RUO
286499
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N9E0
FAM133A
FAM133A Fusion Protein Ag21549
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers