missing translation for 'onlineSavingsMsg'
Learn More

FAM13A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55401-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826865

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to FAM13A. It can recognize all the isoforms of FAM13A.

FAM13A, also named as FAM13A1 and KIAA0914, belongs to the FAM13 family. It is associated with chronic obstructive pulmonary disease.
Specifications

Specifications

FAM13A
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O94988, Q8BGI4
FAM13A, FAM13A1, KIAA0914, Protein FAM13A
Rabbit
IgG
150 μL
-20°C
Primary
10144, 58909
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FAM13A
Liquid
Fam13a
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers