missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM13B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550659
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FAM13B protein.
FAM13B Polyclonal antibody specifically detects FAM13B in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FAM13B | |
Polyclonal | |
Unconjugated | |
FAM13B | |
2610024E20Rik; ARHGAP49; AW060714; AW546153; C5orf5; Fam13b; FAM13B1; family with sequence similarity 13 member B; family with sequence similarity 13, member B; family with sequence similarity 13, member B1; GAP-like protein N61; KHCHP; N61; protein FAM13B; RGD1310484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225358, 51306 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8K2H3, Q9NYF5 | |
FAM13B | |
Synthetic peptide corresponding to a region derived from internal residues of human family with sequence similarity 13, member B | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |