missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM148B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561364
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PCTLESPIRA AAVPRRCAAE SDL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 61%.
FAM148B Polyclonal antibody specifically detects FAM148B in Human samples. It is validated for Immunohistochemistry.Specifications
FAM148B | |
Polyclonal | |
Unconjugated | |
C2CD4B | |
C2 calcium dependent domain containing 4B; C2 calcium-dependent domain containing 4B; C2 calcium-dependent domain-containing protein 4B; C2CD4B; FAM148B; family with sequence similarity 148, member B; NLF2; nuclear localized factor 2; nuclear-localized factor 2; Protein FAM148B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388125 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NLJ0 | |
C2CD4B | |
Recombinant protein corresponding to Human FAM148B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only